(±)-2-Methyl-1-butanol ≥98%
Supplier: Thermo Scientific
|
Warning
|
B21825.0F
B21825.AE
B21825.AP
B21825.0FEA
170
EUR
B21825.0F
B21825.AE
B21825.AP
(±)-2-Methyl-1-butanol ≥98%
(±)-2-Methyl-1-butanol
Purity: 98%
CAS No.: 137-32-6
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 43°(109°F)
Refractive Index: 1.4110
Melting Point:
Formula:
C₅H₁₂O MW: 88,15 g/mol Boiling Pt: 129…131 °C Melting Pt: < –70 °C Density: 0,815 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004743 CAS Number: 137-32-6 EINECS: 205-289-9 UN: 1105 ADR: 3,III Merck Index: 14,6030 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...