(±)-Phenylsuccinic acid 98+%

Supplier: Thermo Fisher Scientific
Warning

130940250 130941000 130945000
ACRO130940250EA 34 EUR
ACRO130940250 ACRO130941000 ACRO130945000
(±)-Phenylsuccinic acid 98+%
(±)-Phenylsuccinic acid
Formula: C₁₀H₁₀O₄
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00004256
CAS Number: 635-51-8
EINECS: 211-238-1

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR