(±)-1-Octen-4-ol ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B20707.06
B20707.14
B20707.06EA
90.7
EUR
B20707.06
B20707.14
(±)-1-Octen-4-ol ≥99%
(±)-1-Octen-4-ol
Purity: 99%
CAS No.: 40575-42-6
Molecular Formula: C8H16O
Formula Weight: 128.22
MDL No.: MFCD00021943
Formula:
C₈H₁₆O MW: 128.21 g/mol Density: 0.83 Storage Temperature: Ambient |
MDL Number:
MFCD00021943 CAS Number: 40575-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...