Guanidinium carbonate

Supplier: Molekula
Warning

Synonyms: Guanidine carbonate salt, Diguanidinium carbonate, Guanidine carbonate

32431272-5KG 32431272-1KG
MOLE32431272-5KGEA 387 EUR
MOLE32431272-5KG MOLE32431272-1KG
Guanidinium carbonate
Guanidinium carbonate
Formula: C₃H₁₂N₆O₃
MW: 180,17 g/mol
Melting Pt: 197 °C
Density: 1,251 g/cm³ (20 °C)
MDL Number: MFCD00013029
CAS Number: 593-85-1
EINECS: 209-813-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR