Guanidinium nitrate

Supplier: Molekula
Warning

Synonyms: Guanidine nitrate

MOLE62000611-1KGEA 108 EUR
MOLE62000611-1KG
Guanidinium nitrate
Guanidinium nitrate
Formula: CH₆N₄O₃
MW: 122,08 g/mol
Melting Pt: 211…214 °C
Density: 1,432 g/cm³ (20 °C)
MDL Number: MFCD00013028
CAS Number: 506-93-4
EINECS: 208-060-1
UN: 1467
ADR: 5.1,III
Merck Index: 13,04578

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR