Guanidinium thiocyanate
Supplier: Roth Carl
|
Warning
|
Synonyms:
Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
0017.1
0017.2
ROTH0017.1EA
45.1
EUR
ROTH0017.1
ROTH0017.2
Guanidinium thiocyanate
Guanidinium thiocyanate
Formula:
CH₅N₃·CHNS MW: 118,16 g/mol Melting Pt: 118 °C Density: 1,29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013027 CAS Number: 593-84-0 EINECS: 209-812-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...