Acetylene-PEG4-maleimide
Supplier: Thermo Scientific
Warning
Synonyms: 3-(N-maleimidyl)-N-(3,6,9,12-tetraoxapentadec-14-yn-1-yl)propionamide
J64859.MB J64859.MC
J64859.MBEA 180 EUR
J64859.MB J64859.MC
Acetylene-PEG4-maleimide
Acetylene-PEG4-maleimide
A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility