Azido-PEG3-carboxyrhodamine 110 conjugate
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Azide-Fluor 488
J65107.X0
J65107.MB
J65107.X0EA
231
EUR
J65107.X0
J65107.MB
Azido-PEG3-carboxyrhodamine 110 conjugate
Azido-PEG3-carboxyrhodamine 110 conjugate
A green fluorescent rhodamine dye for click chemistry reactions and fluorescence microscopy. The PEG spacer improves water solubility and also reduces any steric effect of the dye on the molecule to be labeled.
Formula:
C₂₉H₃₀N₆O₇ MW: 574,59 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD22380746 CAS Number: 1536327-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...