Sodium sulphate decahydrate
Supplier: Roth Carl
X892.1
X892.2
X892.3
ROTHX892.1EA
28.9
EUR
ROTHX892.1
ROTHX892.2
ROTHX892.3
Sodium sulphate decahydrate
Sodium sulphate decahydrate
Formula:
Na₂SO₄·10H₂O MW: 322,2 g/mol Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 32,4 °C Density: 1,464 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149189 CAS Number: 7727-73-3 EINECS: 231-820-9 Merck Index: 13,08755 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...