Calcium sulphate dihydrate
Supplier: Roth Carl
Synonyms:
Plaster of Paris
P741.1
P741.2
ROTHP741.1EA
76.8
EUR
ROTHP741.1
ROTHP741.2
Calcium sulphate dihydrate
Calcium sulphate dihydrate
Formula:
CaSO₄·2H₂O MW: 172,17 g/mol Density: 2,32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149625 CAS Number: 10101-41-4 EINECS: 231-900-3 Merck Index: 13,01711 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...