Tetraethylammonium chloride monohydrate 98+%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Tetrylammonium chloride monohydrate
30818.18
30818.30
30818.18EA
47.9
EUR
30818.18
30818.30
Tetraethylammonium chloride monohydrate 98+%
Tetraethylammonium chloride monohydrate
Purity: 98+%
CAS No.: 68696-18-4
Molecular Formula: (C2H5)4NCl.H2O
Formula Weight: 183.72 (165.71anhy)
Physical Form: Crystalline
Density: 1.0801
Melting Point: ca 110°
Beilstein Reference: 3563247
Merck Reference: 14,9200
MDL No.: MFCD00149992
Phase transfer catalyst
Formula:
C₈H₂₀ClN·1H₂O MW: 183,72 g/mol Melting Pt: ∼ 110 °C Density: 1,0801 Storage Temperature: Ambient |
MDL Number:
MFCD00149992 CAS Number: 68696-18-4 EINECS: 200-267-5 Merck Index: 14,9200 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...