Gadolinium tribromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific
Warning

13208.14 13208.22
13208.14EA 0 EUR
13208.14 13208.22
Gadolinium tribromide hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Gadolinium tribromide hydrate

Purity: 99.99% CAS No.: 30010-20-9 Molecular Formula: GdBr3.xH2O Formula Weight: 396.96(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00049741

Formula: Br₃Gd·H₂O
MW: 414,98 g/mol
Melting Pt: 186 °C
Storage Temperature: Ambient
MDL Number: MFCD00049741
CAS Number: 30010-20-9
EINECS: 237-494-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR