Cerium trichloride heptahydrate 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Cerous chloride heptahydrate, Cerium (III) chloride heptahydrate
211821000
211825000
211820250
ACRO211821000EA
106
EUR
ACRO211821000
ACRO211825000
ACRO211820250
Cerium trichloride heptahydrate 99%
Cerium trichloride heptahydrate
Formula:
CeCl₃·7H₂O MW: 372,58 g/mol Melting Pt: 848 °C Density: 3,97 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149634 CAS Number: 18618-55-8 EINECS: 232-227-8 UN: 1759 ADR: 8,II Merck Index: 12,02041 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Titration Argentometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...