Lutetium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific
11260.03
11260.06
11260.03EA
125
EUR
11260.03
11260.06
Lutetium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Lutetium trichloride hexahydrate
Purity: 99.9%
CAS No.: 15230-79-2
Molecular Formula: LuCl3.6H2O
Formula Weight: 389.42 (281.33anhy)
Physical Form: Crystalline Aggregates
Formula:
Cl₃Lu·6H₂O MW: 389,42 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011099 CAS Number: 15230-79-2 EINECS: 233-240-1 Merck Index: 14,5615 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...