Zinc sulphate heptahydrate

Supplier: Roth Carl
Danger

T884.1 T884.2 T884.3
ROTHT884.1EA 60 EUR
ROTHT884.1 ROTHT884.2 ROTHT884.3
Zinc sulphate heptahydrate
Zinc sulphate heptahydrate
Formula: ZnSO₄·7H₂O
Melting Pt: 100 °C
Density: 1,97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR