Yttrium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific
|
Warning
|
11184.14
11184.22
11184.36
11184.14EA
61.4
EUR
11184.14
11184.22
11184.36
Yttrium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Yttrium (III) chloride hydrate
Purity: 99.9%
CAS No.: 12741-05-8
Molecular Formula: YCl3.xH2O
Formula Weight: 195.26(anhy)
Physical Form: Wet Crystalline Aggregates
Sensitivity: Hygroscopic
Merck Reference: 14,10107
MDL No.: MFCD00149941
Formula:
Cl₃Y·H₂O MW: 213,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149941 CAS Number: 12741-05-8 EINECS: 233-801-0 Merck Index: 14,10107 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...