(+)-Dimethyl-L-tartrate 99%

Supplier: Thermo Fisher Scientific

Synonyms: Dimethyl (2R,3R)-2,3-dihydroxysuccinate, (R,R)-(+)-Tartaric acid dimethyl ester, (R,R)-(+)-Dimethyl tartrate, L(+)-Tartaric acid dimethyl ester, (+)-Dimethyl L-tartrate

172490250 172491000 172495000
ACRO172490250EA 36.8 EUR
ACRO172490250 ACRO172491000 ACRO172495000
(+)-Dimethyl-L-tartrate 99%
(+)-Dimethyl L-tartrate
Formula: C₆H₁₀O₆
Boiling Pt: 157…159 °C (15hPa)
Melting Pt: 48…50 °C
Density: 1,3403 g/cm³ (15 °C)
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00064437
CAS Number: 608-68-4
EINECS: 210-166-8

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +20° to +22° (20°C, 589 nm) (10% in water)
Elemental Analysis 5.22 to 5.65 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR