Potassium-D-gluconate 99%
Supplier: Thermo Scientific
Synonyms:
Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
229320010
229322500
ACRO229320010EA
119
EUR
ACRO229320010
ACRO229322500
Potassium-D-gluconate 99%
Potassium D-gluconate
Formula:
HOH₂C(CH(OH))₄COOK MW: 234,25 g/mol Melting Pt: 178…181 °C Density: 1,73 g/cm³ (20 °C) |
MDL Number:
MFCD00064211 CAS Number: 299-27-4 EINECS: 206-074-2 Merck Index: 12,07796 |
Specification Test Results
Appearance | White to off-white Crystalline or granular powder |
Infrared spectrum | Conforms |
Assay | ≥98.5 % (C6H11KO7) On dried substance |
Loss on drying | ≤3 % (105°C, 4 h) (Vacuum) |
Heavy metals (as Pb) | ≤20 ppm |
Reducing substances | ≤1 % (as Dextrose) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...