L(+)-Potassium sodium tartrate tetrahydrate
Supplier: Roth Carl
Synonyms:
Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate
7998.1
7998.2
7998.3
ROTH7998.1EA
81.2
EUR
ROTH7998.1
ROTH7998.2
ROTH7998.3
L(+)-Potassium sodium tartrate tetrahydrate
L(+)-Potassium sodium tartrate tetrahydrate
Formula:
KOOC(CHOH)₂COONa·4H₂O MW: 282,22 g/mol Melting Pt: 70…80 °C Density: ∼1,767 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150989 CAS Number: 6381-59-5 EINECS: 206-156-8 Merck Index: 13,07755 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...