(R)-(-)-2-Amino-1-propanol 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
D-Alaninol, D(-)-2-Amino-1-propanol, D(-)-Alaninol, (R)-2-Aminopropan-1-ol
400440010
400440050
ACRO400440010EA
46.4
EUR
ACRO400440010
ACRO400440050
(R)-(-)-2-Amino-1-propanol 98%
(R)-(-)-2-Amino-1-propanol
Formula:
C₃H₉NO Boiling Pt: 173 °C (1013 hPa) Density: 0,965 g/cm³ Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064413 CAS Number: 35320-23-1 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4480 to 1.4510 (20°C, 589 nm) |
GC | ≥97.5 % |
Specific optical rotation | -20° to -24° (20°C, 589 nm) (c=2, ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...