L(+)-Leucine
Supplier: Roth Carl
Synonyms:
(S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
3984.1
3984.2
3984.3
ROTH3984.1EA
63.8
EUR
ROTH3984.1
ROTH3984.2
ROTH3984.3
L(+)-Leucine
L(+)-Leucine
Formula:
C₆H₁₃NO₂ MW: 131,17 g/mol Melting Pt: 300 °C Density: 1,165 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002617 CAS Number: 61-90-5 EINECS: 200-522-0 Merck Index: 14,05451 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...