(R)-(-)-2-Amino-3-methyl-1-butanol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 98%
CAS No.: 4276-09-9
Molecular Formula: C5H13NO
Formula Weight: 103.17
Storage Temperature: 2°C to 8°C
Flash Point: 78°(172°F)
Refractive Index: 1.4550
Melting Point: 30-34°
Boiling Point: 189-190°
Beilstein Reference: 1719138
Optical Rotation: -11° (c=10 in water)
MDL No.: MFCD00064297
Formula:
C₅H₁₃NO Boiling Pt: 189…190 °C Melting Pt: 30…34 °C Density: 0,931 Flash Pt: 78 °C (172 °F) |
MDL Number:
MFCD00064297 CAS Number: 4276-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...