(S)-(+)-2-Amino-3-methyl-1-butanol 96%
Supplier: Thermo Scientific
|
Warning
|
181590010
181590050
181590250
ACRO181590010EA
43.8
EUR
ACRO181590010
ACRO181590050
ACRO181590250
(S)-(+)-2-Amino-3-methyl-1-butanol 96%
(S)-(+)-2-Amino-3-methyl-1-butanol
Formula:
C₅H₁₃NO Boiling Pt: 81 °C (8 mmHg) Melting Pt: 30…32 °C Density: 0,92 g/cm³ Flash Pt: 91 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064296 CAS Number: 2026-48-4 EINECS: 217-975-5 |
Specification Test Results
Appearance | Clear colorless to white to light yellow Liquid after melting or solid |
Infrared spectrum | Conforms |
Refractive index | 1.4535 to 1.4565 (20°C, 589 nm) |
GC | ≥95 % |
Specific optical rotation | +15° to +19° (20°C, 589 nm) (c=10, ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...