L(+)-Histidine

Supplier: Roth Carl

Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine

3852.1 3852.2 3852.3
ROTH3852.1EA 0 EUR
ROTH3852.1 ROTH3852.2 ROTH3852.3
L(+)-Histidine
L(+)-Histidine
Formula: C₆H₉N₃O₂
Melting Pt: 272…273 °C
Density: 1,44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064315
CAS Number: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR