D-(+)-Tryptophan ≥99%
Supplier: Thermo Scientific
Synonyms:
H-D-Trp-OH
A18426.06
A18426.14
A18426.22
A18426.06EA
45.8
EUR
A18426.06
A18426.14
A18426.22
D-(+)-Tryptophan ≥99%
D(+)-Tryptophan
Purity: 99%
CAS No.: 153-94-6
Molecular Formula: C11H12N2O2
Formula Weight: 204.23
Melting Point: 273° dec.
Beilstein Reference: 86198
Formula:
C₁₁H₁₂N₂O₂ MW: 204,23 g/mol Melting Pt: 273 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064339 CAS Number: 153-94-6 EINECS: 205-819-9 Merck Index: 14,09797 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...