(R)-(-)-α-Methoxyphenylacetic acid 99%

Supplier: Thermo Scientific
Warning

Synonyms: (R)-(-)-alpha-Methoxyphenylacetic Acid

224470010 224470050
ACRO224470010EA 97.9 EUR
ACRO224470010 ACRO224470050
(R)-(-)-α-Methoxyphenylacetic acid 99%
(R)-(-)-α-Methoxyphenylacetic acid
Formula: C₉H₁₀O₃
MW: 166,18 g/mol
Boiling Pt: 164 - 166 °C (18 -rr)
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00004250
CAS Number: 3966-32-3
EINECS: 223-580-9

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 63 to 68 °C
Specific optical rotation -158° to -162° (20 °C, 589 nm) (c=1, MeOH)
Titration with NaOH 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR