1,7-Octadiene 98.5%

Supplier: Thermo Scientific
Danger

Synonyms: Octa-1,7-diene

129351000 129355000
ACRO129351000EA 109 EUR
ACRO129351000 ACRO129355000
1,7-Octadiene 98.5%
1,7-Octadiene
Formula: C₈H₁₄
MW: 110,2 g/mol
Boiling Pt: 114…121 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0,74 g/cm³
Flash Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008668
CAS Number: 3710-30-3
EINECS: 223-054-9
UN: 2309
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4230 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR