1,7-Octadiyne ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Octa-1,7-diyne
A15829.09
A15829.18
A15829.09EA
96
EUR
A15829.09
A15829.18
1,7-Octadiyne ≥98%
1,7-Octadiyne
Purity: 98%
CAS No.: 871-84-1
Molecular Formula: HC≡C(CH2)4C≡CH
Formula Weight: 106.17
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.4460
Odour: Recognizable
Boiling Point: 135-136°
Beilstein Reference: 1697275
UN No.: 3295
MDL No.: MFCD00008580
Formula:
C₈H₁₀ MW: 106,17 g/mol Boiling Pt: 135…136 °C Melting Pt: –32 °C Density: 0,810 A Flash Pt: 23 °C (73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008580 CAS Number: 871-84-1 EINECS: 212-815-0 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...