Ethyl-5-bromovalerate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A11952.22
A11952.14
A11952.36
A11952.22EA
200
EUR
A11952.22
A11952.14
A11952.36
Ethyl-5-bromovalerate ≥98%
Ethyl 5-bromovalerate
Purity: 98%
CAS No.: 14660-52-7
Molecular Formula: C7H13BrO2
Formula Weight: 209.09
Flash Point: 103°(217°F)
Refractive Index: 1.4580
Boiling Point: 227°
Beilstein Reference: 1752023
MDL No.: MFCD00000266
Formula:
BrH₂C(CH₂)₃COOCH₂CH₃ Boiling Pt: 227 °C Density: 1,306 Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000266 CAS Number: 14660-52-7 EINECS: 238-705-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...