Methyl-5-bromovalerate ≥97%
Supplier: Thermo Fisher Scientific
A19072.06
A19072.14
A19072.22
A19072.06EA
56
EUR
A19072.06
A19072.14
A19072.22
Methyl-5-bromovalerate ≥97%
Methyl 5-bromovalerate
Purity: 97%
CAS No.: 5454-83-1
Molecular Formula: Br(CH2)4CO2CH3
Formula Weight: 195.06
Flash Point: 99°(210°F)
Refractive Index: 1.4630
Boiling Point: 79-80°/4mm
Beilstein Reference: 1749649
MDL No.: MFCD00000265
Formula:
BrH₂C(CH₂)₃COOCH₃ Boiling Pt: 79…80 °C (4 mmHg) Density: 1,363 Flash Pt: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000265 CAS Number: 5454-83-1 EINECS: 226-709-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...