5-Bromovaleric acid ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 5-Bromopentanoic acid

A11765.09 A11765.22 A11765.30
A11765.09EA 23.6 EUR
A11765.09 A11765.22 A11765.30
5-Bromovaleric acid ≥97%
5-Bromovaleric acid

Purity: 97% CAS No.: 2067-33-6 Molecular Formula: Br(CH2)4CO2H Formula Weight: 181.03 Flash Point: 100°(212°F) Melting Point: 38-41° Boiling Point: 142-145°/13mm Beilstein Reference: 1745664 UN No.: 3261 MDL No.: MFCD00004414

Formula: BrH₂C(CH₂)₃COOH
MW: 181,03 g/mol
Boiling Pt: 142…145 °C (13 mmHg)
Melting Pt: 38…41 °C
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004414
CAS Number: 2067-33-6
EINECS: 218-185-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR