Bromoacetic acid 98+%
Supplier: Thermo Scientific
|
Danger
|
106570010
106572500
106570025
106570100
ACRO106570010EA
160
EUR
ACRO106570010
ACRO106572500
ACRO106570025
ACRO106570100
Bromoacetic acid 98+%
Bromoacetic acid
Formula:
BrCH₂COOH Boiling Pt: 208 °C (760 mmHg) Melting Pt: 49…51 °C Density: 1,93 g/cm³ Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002678 CAS Number: 79-08-3 EINECS: 201-175-8 UN: 3425 ADR: 8,II Merck Index: 13,01384 |
Specification Test Results
Appearance | White to yellow Crystalline powder or crystals or flakes or mass and/or chunks |
Infrared spectrum | Conforms |
Melting point | 45 to 49 °C |
Titration with NaOH | 98.0 to 102.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...