Bromoacetic acid ≥98%
Supplier: Thermo Scientific
|
Danger
|
A14403.0B
A14403.0I
A14403.30
A14403.0BEA
106
EUR
A14403.0B
A14403.0I
A14403.30
Bromoacetic acid ≥98%
Bromoacetic acid
Purity: 98+%
CAS No.: 79-08-3
Molecular Formula: C2H3BrO2
Formula Weight: 138.95
Flash Point:
Formula:
BrCH₂COOH Boiling Pt: 207…209 °C Melting Pt: 44…49 °C Density: 1,930 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002678 CAS Number: 79-08-3 EINECS: 201-175-8 UN: 3425 ADR: 8,II Merck Index: 14,1400 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...