3-Chloropropionyl chloride ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
B25698.22
B25698.36
B25698.22EA
50.9
EUR
B25698.22
B25698.36
3-Chloropropionyl chloride ≥97%
3-Chloropropionyl chloride
Purity: 97%
CAS No.: 625-36-5
Molecular Formula: C3H4Cl2O
Formula Weight: 126.97
Flash Point: 59°(138°F)
Refractive Index: 1.4570
Melting Point: -32°
Boiling Point: 143-145°
Beilstein Reference: 635814
UN No.: 3389
MDL No.: MFCD00000747
Formula:
C₃H₄Cl₂O Boiling Pt: 143…145 °C Melting Pt: –32 °C Density: 1,325 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000747 CAS Number: 625-36-5 EINECS: 210-890-4 UN: 3389 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...