Chloroacetamide 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
148410010
148410050
148415000
ACRO148410010EA
102
EUR
ACRO148410010
ACRO148410050
ACRO148415000
Chloroacetamide 98%
Chloroacetamide
Formula:
C₂H₄ClNO Boiling Pt: 256 °C (1013 hPa) Melting Pt: 116…120 °C Density: 0,84 g/cm³ Flash Pt: 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008027 CAS Number: 79-07-2 EINECS: 201-174-2 UN: 2811 ADR: 6.1,III Merck Index: 13,02127 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 116 to 120 °C |
Water | ≤0.2 % |
GC | ≥97.5 % |
Ammonium chloride (NH4Cl) | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...