2',6'-Dichloroacetanilide ≥98%
Supplier: Thermo Scientific
A15195.09
A15195.30
A15195.09EA
0
EUR
A15195.09
A15195.30
2',6'-Dichloroacetanilide ≥98%
2',6'-Dichloroacetanilide
Purity: 98+%
CAS No.: 17700-54-8
Molecular Formula: C8H7Cl2NO
Formula Weight: 204.06
Melting Point: 180-181°
Beilstein Reference: 2096165
Formula:
C₈H₇Cl₂NO Melting Pt: 180…181 °C |
MDL Number:
MFCD00013629 CAS Number: 17700-54-8 EINECS: 241-706-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...