2'-Chloroacetanilide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
N-(2-Chlorophenyl)acetamide
A14711.14
A14711.22
A14711.36
A14711.14EA
33.1
EUR
A14711.14
A14711.22
A14711.36
2'-Chloroacetanilide ≥98%
2'-Chloroacetanilide
Purity: 98+%
CAS No.: 533-17-5
Molecular Formula: C8H8ClNO
Formula Weight: 169.61
Melting Point: 86-88°
Beilstein Reference: 2086967
Merck Reference: 14,2111
MDL No.: MFCD00045169
Formula:
C₈H₈ClNO MW: 169.61 g/mol Boiling Pt: 325 Melting Pt: 86…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045169 CAS Number: 533-17-5 EINECS: 208-555-2 Merck Index: 13,02128 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...