4'-Chloroacetanilide ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Chloroacetanilide, Paracetamol impurity J, N-(4-Chlorophenyl)acetamide
A11214.14
A11214.22
A11214.14EA
78.4
EUR
A11214.14
A11214.22
4'-Chloroacetanilide ≥98%
4'-Chloroacetanilide
Purity: 98+%
CAS No.: 539-03-7
Molecular Formula: C8H8ClNO
Formula Weight: 169.61
Melting Point: 178-180°
Boiling Point: 333°
Merck Reference: 14,2111
MDL No.: MFCD00000612
Formula:
C₈H₈ClNO MW: 169,61 g/mol Boiling Pt: 333 °C Melting Pt: 178…180 °C Density: 1,385 Storage Temperature: Ambient |
MDL Number:
MFCD00000612 CAS Number: 539-03-7 EINECS: 208-707-8 Merck Index: 14,2111 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...