Butyronitrile ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: n-Butyronitrile

L02999.AK L02999.AU
L02999.AKEA 43.5 EUR
L02999.AK L02999.AU
Butyronitrile ≥99%
Butyronitrile

Purity: 99% CAS No.: 109-74-0 Molecular Formula: C4H7N Formula Weight: 69.11 Flash Point: 18°(64°F) Refractive Index: 1.3840 Melting Point: -112° Boiling Point: 115-117° Beilstein Reference: 1361452 Merck Reference: 14,1597 UN No.: 2411 MDL No.: MFCD00001968

Formula: C₄H₇N
MW: 69,11 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –112 °C
Density: 0,792
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001968
CAS Number: 109-74-0
EINECS: 203-700-6
UN: 2411
ADR: 3,II
Merck Index: 14,1597

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR