3-Methoxypropiononitrile 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3-Methoxypropanenitrile, 3-Methoxypropionitrile
257441000
257445000
257440025
ACRO257441000EA
31.7
EUR
ACRO257441000
ACRO257445000
ACRO257440025
3-Methoxypropiononitrile 98%
3-Methoxypropiononitrile
Formula:
C₄H₇NO MW: 85.11 g/mol Boiling Pt: 164…165 °C (760 mmHg) Melting Pt: –62.9 °C Density: 0.938 g/cm³ Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001958 CAS Number: 110-67-8 EINECS: 203-790-7 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4020 to 1.4040 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...