2-Oxoglutaric acid

Supplier: Merck
Danger

Synonyms: 2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid

M-Clarity™ quality level = MQ100

1.05194.0025 1.05194.0100 1.05194.1000 1.05194.9010
1.05194.0025EA 55.9 EUR
1.05194.0025 1.05194.0100 1.05194.1000 1.05194.9010
2-Oxoglutaric acid
2-Oxoglutaric acid
Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
Boiling Pt: 323
Melting Pt: 112…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004165
CAS Number: 328-50-7
EINECS: 206-330-3
Merck Index: 13,05320

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR