Acetyl bromide 98%
Supplier: Thermo Scientific
|
Danger
|
159401000
159405000
ACRO159401000EA
121
EUR
ACRO159401000
ACRO159405000
Acetyl bromide 98%
Acetyl bromide
Formula:
C₂H₃BrO MW: 122,95 g/mol Boiling Pt: 77 °C (1013 hPa) Melting Pt: –96 °C Density: 1,66 g/cm³ Flash Pt: 75 °C |
MDL Number:
MFCD00000114 CAS Number: 506-96-7 EINECS: 208-061-7 UN: 1716 ADR: 8,II Merck Index: 13,00084 |
Specification Test Results
Appearance | Clear colorless to dark yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4480 to 1.4510 (20°C, 589 nm) |
Titration Argentometric | ≥97.5 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤0.001 % |
Heavy metals (as Pb) | ≤0.001 % |
Insoluble matter | ≤0.01 % |
Nitrogen compounds (as N) | ≤0.001 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...