Propionyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A10940.0E A10940.22 A10940.36
A10940.0EEA 214 EUR
A10940.0E A10940.22 A10940.36
Propionyl chloride ≥98%
Propionyl chloride

Purity: 98% CAS No.: 79-03-8 Molecular Formula: C3H5ClO Formula Weight: 92.53 Flash Point: 11°(51°F) Refractive Index: 1.4040 Odour: Unpleasant Melting Point: -94° Boiling Point: 77-79° Beilstein Reference: 385632 Merck Reference: 14,7828 UN No.: 1815 MDL No.: MFCD00000745

Formula: C₃H₅ClO
Boiling Pt: 77…79 °C
Melting Pt: –94 °C
Density: 1,061
Flash Pt: 11 °C (51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3,II
Merck Index: 14,7828

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR