Oxalyl dichloride 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Oxalyl chloride
129610010
129610250
129611000
129610025
ACRO129610010EA
498
EUR
ACRO129610010
ACRO129610250
ACRO129611000
ACRO129610025
Oxalyl dichloride 98%
Oxalyl dichloride
Formula:
C₂Cl₂O₂ MW: 126,93 g/mol Boiling Pt: 63…64 °C (763 mmHg) Melting Pt: –12 °C Density: 1,478 g/cm³ Flash Pt: >100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000704 CAS Number: 79-37-8 EINECS: 201-200-2 UN: 3130 ADR: 4.3,I Merck Index: 12,07046 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4285 to 1.4305 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...