N,N-Diethylacetamide for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
N-Acetyldiethylamine
M-Clarity™ quality level = MQ200
8.03214.0025
8.03214.0100
8.03214.0025EA
36.7
EUR
8.03214.0025
8.03214.0100
N,N-Diethylacetamide for synthesis, Sigma-Aldrich®
N,N-Diethylacetamide
Formula:
C₆H₁₃NO MW: 115,18 g/mol Boiling Pt: 182 °C (1013 hPa) Density: 1,02 g/cm³ (20 °C) Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009047 CAS Number: 685-91-6 EINECS: 211-685-2 |
Specification Test Results
Solubility | 53.5 g/l (25 °C) |
Molar Mass | 115.17 g/mol |
Boiling Point | 182 - 186 °C (1013 hPa) |
Vapor Pressure | 1.2 hPa (25 °C) |
Flash Point | 76 °C |
Refractive Index | 1.4401 (20 °C, 589 nm) |
Density | 0.91 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...