N,N-Dibutylformamide 99+%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Formdibutylamide, Formic acid dibutylamide
112980100
112985000
ACRO112980100EA
49.4
EUR
ACRO112980100
ACRO112985000
N,N-Dibutylformamide 99+%
N,N-Dibutylformamide
Formula:
C₉H₁₉NO MW: 157.26 g/mol Boiling Pt: 120 °C (15 mmHg) Melting Pt: -30 Density: 0.86 g/cm³ Flash Pt: 100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003289 CAS Number: 761-65-9 EINECS: 212-090-0 UN: 2810 ADR: 6.1 III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4430 to 1.4450 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥99.0 % |
HPLC | 48.0 to 52.0 % Hydroquinone |
Sulfate (SO4) | ≤200 ppm |
Iron (Fe) | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...