N,N-Dibutylformamide ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: Formdibutylamide, Formic acid dibutylamide

L12755.22 L12755.36 L12755.14
L12755.22EA 96 EUR
L12755.22 L12755.36 L12755.14
N,N-Dibutylformamide ≥99%
N,N-Dibutylformamide

Purity: 99% CAS No.: 761-65-9 Molecular Formula: C9H19NO Formula Weight: 157.26 Flash Point: 115°(239°F) Refractive Index: 1.4445 Melting Point: -30° Boiling Point: 119-120°/15mm Beilstein Reference: 636100 MDL No.: MFCD00003289

Formula: C₉H₁₉NO
MW: 157,26 g/mol
Boiling Pt: 119…120 °C (15 mmHg)
Melting Pt: < –30 °C
Density: 0,864
Flash Pt: 115 °C (239 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003289
CAS Number: 761-65-9
EINECS: 212-090-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR