4'-Nitroacetanilide for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: N-(4-Nitrophenyl)acetamide

M-Clarity™ quality level = MQ200

8.20880.0025 8.20880.0100
8.20880.0025EA 47.7 EUR
8.20880.0025 8.20880.0100
4'-Nitroacetanilide for synthesis, Sigma-Aldrich®
4'-Nitroacetanilide
Formula: C₈H₈N₂O₃
MW: 180,16 g/mol
Boiling Pt: 409 °C (1013 hPa)
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00007303
CAS Number: 104-04-1
EINECS: 203-169-0
Merck Index: 12,06677

Order Now

Specification Test Results

Solubility 2.2 g/l (25 °C)
Melting Point 212 - 215 °C
Molar Mass 180.16 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR