Butyl laurate 99

Supplier: Thermo Fisher Scientific

Synonyms: Butyl dodecanoate, Dodecylic acid butyl ester, Dodecanoic acid butyl ester, Lauric acid butyl ester, n-Butyl dodecanoate

42024.AC 42024.AE 42024.AP
42024.ACEA 33.4 EUR
42024.AC 42024.AE 42024.AP
Butyl laurate 99
Butyl laurate

Purity: 99+% CAS No.: 106-18-3 Molecular Formula: CH3(CH2)10COO(CH2)3CH3 Formula Weight: 256.4 Flash Point: 110°(230°F) Physical Form: Liquid Density: 0.86 Refractive Index: 1.435 Melting Point: -10° Boiling Point: 166° Beilstein Reference: 1776360 MDL No.: MFCD00042870

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 166 °C
Melting Pt: –10 °C
Density: 0.86
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042870
CAS Number: 106-18-3
EINECS: 203-370-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR