Ethyl stearate 97%
Supplier: Thermo Scientific
Synonyms:
Stearic acid ethyl ester, Ethyl octadecanoate
207480050
207480250
ACRO207480050EA
39
EUR
ACRO207480050
ACRO207480250
Ethyl stearate 97%
Ethyl stearate
Formula:
C₂₀H₄₀O₂ MW: 312,54 g/mol Boiling Pt: 213…215 °C (15 mmHg) Melting Pt: 30…39 °C Density: 0,8973 g/cm³ (25 °C) Flash Pt: >112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009006 CAS Number: 111-61-5 EINECS: 203-887-4 |
Specification Test Results
Appearance | White to light yellow Powder or low melting crystalline mass or liquid |
Infrared spectrum | Conforms |
Melting point | 30 to 39 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...